Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Fluoro-2,3-dimethoxybenzene
1-Fluoro-2,3-dimethoxybenzene 95%
1-Fluoro-2,3-dimethoxybenzene 95%
APOSPC906944-1G
:
Apollo Scientific
:
1-Fluoro-2,3-dimethoxybenzene 95%
APOSPC906944-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
156.154 g/mol
:
Ambient
:
394-64-9
:
C₈H₉FO₂
:
MFCD03411451