Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Dichloro-3-fluorobenzaldehyde
2,6-Dichloro-3-fluorobenzaldehyde 95%
2,6-Dichloro-3-fluorobenzaldehyde 95%
2,6-Dichloro-3-fluorobenzaldehyde 95%
Supplier:
Apollo Scientific
Certificates
Expand image
2,6-Dichloro-3-fluorobenzaldehyde 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
178813-77-9
:
Formula:
C₇H₃Cl₂FO
MDL Number:
MFCD09835189
:
193.003 g/mol
Melting Pt:
63…65 °C
Storage Temperature:
Ambient