Order Entry
Export
ContactUsLinkComponent
3,5-Dimethoxy-2-fluoroaniline 95%
3,5-Dimethoxy-2-fluoroaniline 95%
Catalog # APOSPC906804-1G
Supplier:  Apollo Scientific
CAS Number:  
3,5-Dimethoxy-2-fluoroaniline 95%
Catalog # APOSPC906804-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :171.169 g/mol
Storage Temperature:Ambient
CAS Number:651734-61-1
Formula:C₈H₁₀FNO₂
MDL Number:MFCD20696760