Order Entry
Export
ContactUsLinkComponent
2-Chloro-3,4-difluoroaniline 95%
2-Chloro-3,4-difluoroaniline 95%
Catalog # APOSPC906534-25G
Supplier:  Apollo Scientific
CAS Number:  
2-Chloro-3,4-difluoroaniline 95%
Catalog # APOSPC906534-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :163.552 g/mol
Storage Temperature:Ambient
CAS Number:36556-48-6
Formula:C₆H₄ClF₂N
MDL Number:MFCD11044823