2-Fluorobenzene-1,4-diol 95%
New Product
Supplier: Apollo Scientific
Jump to:
About this item
- CAS Number:55660-73-6
- Formula:C₆H₅FO₂
- MDL Number:MFCD00061106
- Molecular Weight:128.101 g/mol
- Storage Temperature:Ambient
5 Options Available Below
Product Details & Documents
- CAS Number:55660-73-6
- Formula:C₆H₅FO₂
- MDL Number:MFCD00061106
- Molecular Weight:128.101 g/mol
- Storage Temperature:Ambient
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



