Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(Trifluoroacetyl)cyclohexanone
2-(Trifluoroacetyl)cyclohexanone
2-(Trifluoroacetyl)cyclohexanone
Irritant
2-(Trifluoroacetyl)cyclohexanone
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2-(Trifluoroacetyl)cyclohexanone
Supplier:
Apollo Scientific
About this item
CAS Number:
387-89-3
:
Formula:
C₈H₉F₃O₂
MDL Number:
MFCD07779548
:
194.151 g/mol
:
GHS07
Storage Temperature:
Ambient