Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,5-Difluoro-4-methylaniline
2,5-Difluoro-4-methylaniline 97%
2,5-Difluoro-4-methylaniline 97%
APOSPC904700-1G
:
Apollo Scientific
:
2,5-Difluoro-4-methylaniline 97%
APOSPC904700-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
143.134 g/mol
:
GHS07
:
Ambient
:
878285-14-4
:
C₇H₇F₂N
:
MFCD11519027