
3,6-Dichloro-4-fluoropyridazine
Catalog # APOSPC904584-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:166.969 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:92920-33-7
Formula:C₄HCl₂FN₂
MDL Number:MFCD21648467