Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-(Trifluoromethyl)-1H-pyrazolo[3,4-b]pyridine
3-(Trifluoromethyl)-1H-pyrazolo[3,4-b]pyridine 95%
3-(Trifluoromethyl)-1H-pyrazolo[3,4-b]pyridine 95%
3-(Trifluoromethyl)-1H-pyrazolo[3,4-b]pyridine 95%
Supplier:
Apollo Scientific
Certificates
Expand image
3-(Trifluoromethyl)-1H-pyrazolo[3,4-b]pyridine 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
956010-87-0
:
Formula:
C₇H₄F₃N₃
MDL Number:
MFCD13178059
:
187.122 g/mol
Storage Temperature:
Ambient