Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Dibromo-5-fluoropyrimidine
2,4-Dibromo-5-fluoropyrimidine 95+%
2,4-Dibromo-5-fluoropyrimidine 95+%
:
Apollo Scientific
2,4-Dibromo-5-fluoropyrimidine 95+%
:
Apollo Scientific
:
1372096-33-7
:
:
C₄HBr₂FN₂
:
MFCD21647860
:
255.871 g/mol
:
GHS07
:
Refrigerator