Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,4-Difluoro-2-methylbenzaldehyde
3,4-Difluoro-2-methylbenzaldehyde 95%
3,4-Difluoro-2-methylbenzaldehyde 95%
3,4-Difluoro-2-methylbenzaldehyde 95%
APOSPC903673-1G
:
Apollo Scientific
:
3,4-Difluoro-2-methylbenzaldehyde 95%
APOSPC903673-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
156.129 g/mol
:
Ambient
:
847502-84-5
:
C₈H₆F₂O
:
MFCD07777151