Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(E)-[(2,5-Difluorophenyl)methylidene]hydrazine
(E)-[(2,5-Difluorophenyl)methylidene]hydrazine 95%
(E)-[(2,5-Difluorophenyl)methylidene]hydrazine 95%
APOSPC902909-1G
:
Apollo Scientific
:
(E)-[(2,5-Difluorophenyl)methylidene]hydrazine 95%
APOSPC902909-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
156.133 g/mol
:
Ambient
:
C₇H₆F₂N₂
:
MFCD25953892