Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chloro-1,3-difluoro-5-nitrobenzene
2-Chloro-1,3-difluoro-5-nitrobenzene 97%
2-Chloro-1,3-difluoro-5-nitrobenzene 97%
2-Chloro-1,3-difluoro-5-nitrobenzene 97%
Supplier:
Apollo Scientific
Certificates
Expand image
2-Chloro-1,3-difluoro-5-nitrobenzene 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
3828-41-9
:
Formula:
C₆H₂ClF₂NO₂
MDL Number:
MFCD09833603
:
193.535 g/mol
Storage Temperature:
Ambient