Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-(2-Fluorophenyl)-2,5-dimethylpyrrole
1-(2-Fluorophenyl)-2,5-dimethylpyrrole 96%
1-(2-Fluorophenyl)-2,5-dimethylpyrrole 96%
1-(2-Fluorophenyl)-2,5-dimethylpyrrole 96%
Supplier:
Apollo Scientific
Certificates
Expand image
1-(2-Fluorophenyl)-2,5-dimethylpyrrole 96%
Supplier:
Apollo Scientific
About this item
CAS Number:
146135-20-8
:
Formula:
C₁₂H₁₂FN
MDL Number:
MFCD07429174
:
189.229 g/mol
Storage Temperature:
Ambient