Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-(2,4-Difluorophenyl)pyrrolidine
1-(2,4-Difluorophenyl)pyrrolidine 95%
1-(2,4-Difluorophenyl)pyrrolidine 95%
1-(2,4-Difluorophenyl)pyrrolidine 95%
Supplier:
Apollo Scientific
Certificates
Expand image
1-(2,4-Difluorophenyl)pyrrolidine 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
1334499-91-0
:
Formula:
C₁₀H₁₁F₂N
MDL Number:
MFCD20231457
:
183.198 g/mol
Storage Temperature:
Ambient