Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Fluoro-2,4-dimethoxyphenylboronic acid
3-Fluoro-2,4-dimethoxyphenylboronic acid 98%
3-Fluoro-2,4-dimethoxyphenylboronic acid 98%
3-Fluoro-2,4-dimethoxyphenylboronic acid 98%
Supplier:
Apollo Scientific
Certificates
Expand image
3-Fluoro-2,4-dimethoxyphenylboronic acid 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
949146-39-8
:
Formula:
C₈H₁₀BFO₄
MDL Number:
MFCD23381042
:
199.972 g/mol
Storage Temperature:
Ambient