Order Entry
Export
ContactUsLinkComponent
1-Acetyl-3,5-bis(trifluoromethyl)-1H-pyrazole
Irritant
1-Acetyl-3,5-bis(trifluoromethyl)-1H-pyrazole
Catalog # APOSPC9009-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-Acetyl-3,5-bis(trifluoromethyl)-1H-pyrazole
Catalog # APOSPC9009-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :246.11 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:244187-01-7
Formula:C₇H₄F₆N₂O
MDL Number:MFCD00153613