
4-Bromo-2-(trifluoromethyl)-1,3-benzoxazole
Catalog # APOSPC8917-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:266.015 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:217326-71-1
Formula:C₈H₃BrF₃NO
MDL Number:MFCD17010236