Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,4-Bis(bromodifluoromethyl)benzene
1,4-Bis(bromodifluoromethyl)benzene
1,4-Bis(bromodifluoromethyl)benzene
:
Apollo Scientific
1,4-Bis(bromodifluoromethyl)benzene
:
Apollo Scientific
:
651-12-7
:
:
C₈H₄Br₂F₄
:
MFCD02270920
:
335.919 g/mol
:
107…108 °C (29 mm Hg)
:
1.95 g/cm³ (25 °C)
:
>110 °C
:
GHS07
:
Ambient