Order Entry
Export
ContactUsLinkComponent
2,4-Dichloro-6-(3-fluorophenoxy)-1,3,5-triazine
Irritant
2,4-Dichloro-6-(3-fluorophenoxy)-1,3,5-triazine
Catalog # APOSPC8098-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,4-Dichloro-6-(3-fluorophenoxy)-1,3,5-triazine
Catalog # APOSPC8098-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :260.052 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:112748-45-5
Formula:C₉H₄Cl₂FN₃O
MDL Number:MFCD05154988