Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Bromo-1,3-dimethyl-5-fluorobenzene
2-Bromo-1,3-dimethyl-5-fluorobenzene 98%
2-Bromo-1,3-dimethyl-5-fluorobenzene 98%
:
Apollo Scientific
2-Bromo-1,3-dimethyl-5-fluorobenzene 98%
:
Apollo Scientific
:
14659-58-6
:
:
C₈H₈BrF
:
MFCD09864702
:
203.052 g/mol
:
GHS07
:
Ambient