Order Entry
Export
ContactUsLinkComponent
1,4-Difluoro-2,3-dimethylbenzene 96%
Flammable
Irritant
1,4-Difluoro-2,3-dimethylbenzene 96%
Catalog # APOSPC7866-5G
Supplier:  Apollo Scientific
CAS Number:  
Flammable
Irritant
1,4-Difluoro-2,3-dimethylbenzene 96%
Catalog # APOSPC7866-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :142.146 g/mol
Boiling Pt:155 °C
Density:1.09 g/cm³ (25 °C)
Flash Pt:36 °C
UN:1993
ADR:3,III
 :GHS02
 :GHS07
Storage Temperature:Ambient
CAS Number:1736-90-9
Formula:C₈H₈F₂
MDL Number:MFCD04973769