
4,6-Difluoro-1H-benzimidazole
Catalog # APOSPC7239-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:154.117 g/mol
Melting Pt:235…238 °C
:GHS07
Storage Temperature:Ambient
CAS Number:2208-24-4
Formula:C₇H₄F₂N₂
MDL Number:MFCD03425145