Order Entry
Export
Orders LinkContactUsLinkComponent
1,1-Difluorodimethylether

1,1-Difluorodimethylether

Bottle
Catalog # APOSPC7155-25G
Compressed Gas
Irritant
New Product
Supplier:   Apollo Scientific
1,1-Difluorodimethylether
Catalog # APOSPC7155-25G
Supplier:  Apollo Scientific
New Product
CAS Number:  359-15-9
1,1-Difluorodimethylether

Bottle

CAS Number:  359-15-9
Price
682,00 €
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:359-15-9
  • Formula:C₂H₄F₂O
  • MDL Number:MFCD07784223
  • Molecular Weight:82.049 g/mol
  • Boiling Pt:–4 °C (740 mm Hg)
  • Storage Temperature:Ambient
  • UN:3161
  • ADR:2.1,
Specifications

Specifications

  • Pack type:
    Bottle

Specifications