Order Entry
Export
ContactUsLinkComponent
 
4,4-Bis(trifluoromethyl)oxetan-2-one
Catalog # APOSPC7002-1G
Supplier:  Apollo Scientific
CAS Number:  
4,4-Bis(trifluoromethyl)oxetan-2-one
Catalog # APOSPC7002-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :208.059 g/mol
Boiling Pt:94.5…95 °C (775 mm Hg)
Density:1.6561 g/cm³ (25 °C)
Storage Temperature:Ambient
CAS Number:1718-33-8
Formula:C₅H₂F₆O₂
MDL Number:MFCD01709493