Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(Trifluoromethoxy)benzene-1,3-diamine
4-(Trifluoromethoxy)benzene-1,3-diamine
4-(Trifluoromethoxy)benzene-1,3-diamine
APOSPC6792-1G
:
Apollo Scientific
:
4-(Trifluoromethoxy)benzene-1,3-diamine
APOSPC6792-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
192.138 g/mol
:
Ambient
:
2811
:
6.1,III
:
GHS06
:
873055-90-4
:
C₇H₇F₃N₂O
:
MFCD04972990