
11-(Perfluorohex-1-yl)undecan-1-ol
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:134052-01-0
:
Formula:C₁₇H₂₃F₁₃O
MDL Number:MFCD00236706
:490.343 g/mol
Boiling Pt:110…115 °C (1 mm Hg)
Melting Pt:49…51 °C
:GHS07
Storage Temperature:Ambient