

3-(2H-Tetrafluoroethoxy)-1,2-propenoxide 97%
Catalog # APOSPC6006G-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:174.094 g/mol
Boiling Pt:142…144 °C
Density:1.38 g/cm³ (25 °C)
Flash Pt:52 °C
Storage Temperature:Ambient
EINECS:287-734-7
UN:3271
ADR:3,III
:GHS02
:GHS07
CAS Number:85567-21-1
Formula:C₅H₆F₄O₂
MDL Number:MFCD00043993