Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,5-Bis(trifluoromethyl)-4-chloroaniline
3,5-Bis(trifluoromethyl)-4-chloroaniline 97%
3,5-Bis(trifluoromethyl)-4-chloroaniline 97%
:
Apollo Scientific
3,5-Bis(trifluoromethyl)-4-chloroaniline 97%
:
Apollo Scientific
:
948014-37-7
:
:
C₈H₄ClF₆N
:
MFCD09763619
:
6.1,III
:
263.567 g/mol
:
2810
:
GHS07
:
Ambient