Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,5-(Dimethyl)perfluorocyclooctane
1,5-(Dimethyl)perfluorocyclooctane
1,5-(Dimethyl)perfluorocyclooctane
1,5-(Dimethyl)perfluorocyclooctane
Supplier:
Apollo Scientific
Certificates
Expand image
1,5-(Dimethyl)perfluorocyclooctane
Supplier:
Apollo Scientific
About this item
CAS Number:
948014-28-6
:
Formula:
C₁₀H₆F₁₄
MDL Number:
MFCD12026454
:
392.132 g/mol
Storage Temperature:
Ambient