Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,2-Dichloro-4-iodoperfluorobutane
1,2-Dichloro-4-iodoperfluorobutane
1,2-Dichloro-4-iodoperfluorobutane
APOSPC5827-5G
:
Apollo Scientific
:
1,2-Dichloro-4-iodoperfluorobutane
APOSPC5827-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
378.842 g/mol
:
GHS07
:
Ambient
:
678-13-7
:
C₄Cl₂F₇I
:
MFCD00042241