
1H-Perfluorohexane-1,1-diol
Catalog # APOSPC5791-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:316.069 g/mol
Melting Pt:70…72 °C
:GHS07
Storage Temperature:Ambient
CAS Number:904037-24-7
Formula:C₆H₃F₁₁O₂
MDL Number:MFCD00437976