Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,4-Difluoro-5-nitrobenzonitrile
3,4-Difluoro-5-nitrobenzonitrile 95%
3,4-Difluoro-5-nitrobenzonitrile 95%
3,4-Difluoro-5-nitrobenzonitrile 95%
APOSPC56908-250MG
:
Apollo Scientific
:
3,4-Difluoro-5-nitrobenzonitrile 95%
APOSPC56908-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
183.112 g/mol
:
GHS07
:
Refrigerator
:
1119454-07-7
:
C₈H₃F₂NO₂
:
MFCD11110377