
8-Iodo-6-(trifluoromethyl)imidazo[1,5-a]pyrimidine 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2151834-40-9
:
Formula:C₇H₃F₃IN₃
MDL Number:MFCD31664911
:313.018 g/mol
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS06