Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Bromo-2,5-difluorobenzeneboronic acid
4-Bromo-2,5-difluorobenzeneboronic acid 97%
4-Bromo-2,5-difluorobenzeneboronic acid 97%
Irritant
4-Bromo-2,5-difluorobenzeneboronic acid 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4-Bromo-2,5-difluorobenzeneboronic acid 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
1106676-82-7
:
Formula:
C₆H₄BBrF₂O₂
MDL Number:
MFCD07644467
:
236.807 g/mol
:
GHS07
Storage Temperature:
Refrigerator