0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Methyl 2,3-difluoro-4-hydroxybenzoate
Methyl 2,3-difluoro-4-hydroxybenzoate
Methyl 2,3-difluoro-4-hydroxybenzoate
:
Apollo Scientific
Methyl 2,3-difluoro-4-hydroxybenzoate
:
Apollo Scientific
:
219685-84-4
:
:
C₈H₆F₂O₃
:
MFCD11042793
:
188.128 g/mol
:
GHS07
:
Refrigerator