Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,2-Difluoropropanenitrile
2,2-Difluoropropanenitrile
2,2-Difluoropropanenitrile
:
Apollo Scientific
2,2-Difluoropropanenitrile
:
Apollo Scientific
:
49781-49-9
:
:
C₃H₃F₂N
:
MFCD07780127
:
91.059 g/mol
:
Refrigerator
:
3276
:
6.1,III
:
GHS06