Order Entry
Export
ContactUsLinkComponent
4,6-Difluoroindan-1-one 98%
Irritant
4,6-Difluoroindan-1-one 98%
Catalog # APOSPC5481-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4,6-Difluoroindan-1-one 98%
Catalog # APOSPC5481-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :168.14 g/mol
Melting Pt:98…99 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:162548-73-4
Formula:C₉H₆F₂O
MDL Number:MFCD04972815