Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,2-Difluorobenzo[d][1,3]dioxol-5-ol
2,2-Difluorobenzo[d][1,3]dioxol-5-ol ≥98%
2,2-Difluorobenzo[d][1,3]dioxol-5-ol ≥98%
2,2-Difluorobenzo[d][1,3]dioxol-5-ol ≥98%
Supplier:
Apollo Scientific
Certificates
Expand image
2,2-Difluorobenzo[d][1,3]dioxol-5-ol ≥98%
Supplier:
Apollo Scientific
About this item
CAS Number:
1211539-82-0
:
Formula:
C₇H₄F₂O₃
MDL Number:
MFCD22414499
:
174.102 g/mol
Storage Temperature:
Ambient