2-Chloro-4-(2,2,2-trifluoroethoxy)pyrimidine > 97%
Catalog # APOSPC540018-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:212.557 g/mol
Storage Temperature:Freezer
CAS Number:935252-67-8
Formula:C₆H₄ClF₃N₂O
MDL Number:MFCD16712374