Order Entry
Export
ContactUsLinkComponent
3,4-Difluoro-2-formylphenylboronic acid ≥98%
3,4-Difluoro-2-formylphenylboronic acid ≥98%
Catalog # APOSPC540012-250MG
Supplier:  Apollo Scientific
CAS Number:  
3,4-Difluoro-2-formylphenylboronic acid ≥98%
Catalog # APOSPC540012-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :185.921 g/mol
Storage Temperature:Refrigerator
CAS Number:1451392-91-8
Formula:C₇H₅BF₂O₃
MDL Number:MFCD23713092