Order Entry
Export
ContactUsLinkComponent
2-Chloro-5,6-difluoronicotinonitrile ≥95%
2-Chloro-5,6-difluoronicotinonitrile ≥95%
Catalog # APOSPC540005-250MG
Supplier:  Apollo Scientific
CAS Number:  
2-Chloro-5,6-difluoronicotinonitrile ≥95%
Catalog # APOSPC540005-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :174.535 g/mol
Storage Temperature:Refrigerator
CAS Number:1309371-71-8
Formula:C₆HClF₂N₂
MDL Number:MFCD27996746