Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3'-Chloro-4'-iodo-2,2,2-trifluoroacetophenone
3'-Chloro-4'-iodo-2,2,2-trifluoroacetophenone 95%
3'-Chloro-4'-iodo-2,2,2-trifluoroacetophenone 95%
Irritant
3'-Chloro-4'-iodo-2,2,2-trifluoroacetophenone 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3'-Chloro-4'-iodo-2,2,2-trifluoroacetophenone 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
2352970-26-2
:
Formula:
C₈H₃ClF₃IO
MDL Number:
MFCD32200319
:
334.461 g/mol
:
GHS07
Storage Temperature:
Ambient