Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,5-Difluoro-4-iodobenzotrifluoride
3,5-Difluoro-4-iodobenzotrifluoride
3,5-Difluoro-4-iodobenzotrifluoride
Irritant
3,5-Difluoro-4-iodobenzotrifluoride
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3,5-Difluoro-4-iodobenzotrifluoride
Supplier:
Apollo Scientific
About this item
CAS Number:
2149601-44-3
:
Formula:
C₇H₂F₅I
MDL Number:
MFCD30729941
:
307.987 g/mol
:
GHS07
Storage Temperature:
Ambient