Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,6-Dichloro-2-fluoropyrimidine
4,6-Dichloro-2-fluoropyrimidine 95+%
4,6-Dichloro-2-fluoropyrimidine 95+%
Irritant
4,6-Dichloro-2-fluoropyrimidine 95+%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4,6-Dichloro-2-fluoropyrimidine 95+%
Supplier:
Apollo Scientific
About this item
CAS Number:
3824-45-1
:
Formula:
C₄HCl₂FN₂
MDL Number:
MFCD27939250
:
166.969 g/mol
:
GHS07
Storage Temperature:
Refrigerator