Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,5-Difluoro-2-iodotoluene
4,5-Difluoro-2-iodotoluene
4,5-Difluoro-2-iodotoluene
:
Apollo Scientific
4,5-Difluoro-2-iodotoluene
:
Apollo Scientific
:
1208077-21-7
:
:
C₇H₅F₂I
:
MFCD12547764
:
254.016 g/mol
:
GHS07
:
Refrigerator