Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,5-Bis(trifluoromethyl)pyridine
2,5-Bis(trifluoromethyl)pyridine 99%
2,5-Bis(trifluoromethyl)pyridine 99%
Irritant
2,5-Bis(trifluoromethyl)pyridine 99%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2,5-Bis(trifluoromethyl)pyridine 99%
Supplier:
Apollo Scientific
About this item
CAS Number:
20857-44-7
:
Formula:
C₇H₃F₆N
MDL Number:
MFCD00128906
:
215.096 g/mol
:
GHS07
Storage Temperature:
Ambient