Order Entry
Export
Orders LinkContactUsLinkComponent
4-(1,1-Difluoroethyl)benzaldehyde

4-(1,1-Difluoroethyl)benzaldehyde

Irritant
Supplier:   Apollo Scientific
4-(1,1-Difluoroethyl)benzaldehyde
Supplier:  Apollo Scientific
Certificates
4-(1,1-Difluoroethyl)benzaldehyde

2 Options Available Below

Product Details & Documents
  • CAS Number:55805-22-6
  • Formula:C₉H₈F₂O
  • MDL Number:MFCD17015610
  • Molecular Weight:170.156 g/mol
  • Storage Temperature:Refrigerator
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 312,00 €
    312,00 €
    1 * 250 mg
       312,00 €

  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 738,00 €
    738,00 €
    1 * 1 g
       738,00 €