Order Entry
Export
ContactUsLinkComponent
6-Fluoroimidazo[1,2-b]-pyridazine ≥98%
6-Fluoroimidazo[1,2-b]-pyridazine ≥98%
Catalog # APOSPC53315-1G
Supplier:  Apollo Scientific
CAS Number:  
6-Fluoroimidazo[1,2-b]-pyridazine ≥98%
Catalog # APOSPC53315-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :137.114 g/mol
Storage Temperature:Refrigerator
CAS Number:113501-27-2
Formula:C₆H₄FN₃
MDL Number:MFCD11044764