Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-(Trifluoromethyl)-1H-imidazo[4,5-b]pyridine
5-(Trifluoromethyl)-1H-imidazo[4,5-b]pyridine
5-(Trifluoromethyl)-1H-imidazo[4,5-b]pyridine
Irritant
5-(Trifluoromethyl)-1H-imidazo[4,5-b]pyridine
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
5-(Trifluoromethyl)-1H-imidazo[4,5-b]pyridine
Supplier:
Apollo Scientific
About this item
CAS Number:
617678-32-7
:
Formula:
C₇H₄F₃N₃
MDL Number:
MFCD13193496
:
187.122 g/mol
:
GHS07
Storage Temperature:
Ambient